missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, ACS Reagent, ≥99.4%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -90°C |
| Density | 0.81g/mL at 25°C |
| Boiling Point | 116°C |
| Quantity | 500 mL |
| Explosion Limit | 0.112 |
| Flash Point | 35°C (95°F) |
| Ignition Point | 649°F |
| Infrared Spectrum | Conforms to structure |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction