missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, ACS Reagent, ≥99.4%, Solstice

Product Code. 15655060
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15655060 4 x 2.5 L Glass Bottle Pack of 4
15665060 500 mL Glass Bottle 500mL
15635060 1 L Glass Bottle 1 L
15645060 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15655060 Supplier Honeywell-Riedel-de Haen Supplier No. 3604654X2.5L
  • ONLINE EXCLUSIVE PRICE
    £441.00
    £332.05 / Pack of 4
    Save £108.95
    24% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.4%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Density 0.81g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.112
Flash Point 35°C (95°F)
Ignition Point 649°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Boiling Point 116°C
Formula Weight 74.12g/mol
Percent Purity ≥99.4%
Chemical Name or Material 1-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.