missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 98%, AcroSeal™, Thermo Scientific™

Product Code. 10412098 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412098 100 mL AcroSeal™ Glass bottle 100mL
10747518 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412098 Supplier Thermo Scientific Acros Supplier No. 430871000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -116.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 98.0°C
Flash Point 0.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula CH3(CH2)3SH
Refractive Index 1.4420 to 1.4440
Quantity 100 mL
Merck Index 15,158
Specific Gravity 0.84
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 90.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.