missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 98%, Thermo Scientific Chemicals

Catalog Number 11460580
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
109-79-5
C4H10S
90.184
MFCD00004905
WQAQPCDUOCURKW-UHFFFAOYSA-N
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
8012
butane-1-thiol
CCCCS
This item is not returnable. View return policy
109-79-5
C4H10S
90.184
MFCD00004905
WQAQPCDUOCURKW-UHFFFAOYSA-N
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
8012
butane-1-thiol
CCCCS

1-Butanethiol is used as a model gaseous analyte for fast-responding membrane-free amperometric gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -116°C
Color Colorless
Density 0.842 g/mL
Boiling Point 98°C
Flash Point 2°C (36°F)
Odor Stench
Linear Formula CH3(CH2)3SH
Refractive Index 1.4425
Quantity 25 mL
UN Number UN2347
Beilstein 1730908
Sensitivity Air Sensitive
Merck Index 14,1577
Solubility Information Soluble in water (0.59g/L at 20°C).
Formula Weight 90.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'