missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonyl chloride, 98%

Product Code. 15750869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15750869 100 g Glass bottle 100g
10299350 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15750869 Supplier Thermo Scientific Acros Supplier No. 403321000
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2386-60-9 | C4H9ClO2S | 156.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.63
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or Orange
Density 1.2000g/mL
Boiling Point 96.0°C (21.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3SO2Cl
Refractive Index 1.4540 to 1.4565
Quantity 100 g
Specific Gravity 1.2
Solubility Information Solubility in water: decomposes.
Formula Weight 156.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanesulfonyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.