missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonic acid, sodium salt, 98%

Product Code. 10420194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420194 100 g Glass bottle 100g
10780711 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420194 Supplier Thermo Scientific Acros Supplier No. 206101000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2386-54-1 | C4H9NaO3S | 160.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
PubChem CID 4096517
IUPAC Name sodium;butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 2% max. (1 g, 105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)3SO3Na
Quantity 100 g
Beilstein 04, 8
Solubility Information (10% in water) Clear to slightly hazy colorless
Formula Weight 160.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Butanesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 1-Butanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.