missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%

Product Code. 10184510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184510 100 g Glass bottle 100g
10582811 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10184510 Supplier Thermo Scientific Acros Supplier No. 224011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-67-4 | C11H23Br | 235.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-67-4
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H23Br
Linear Formula CH3(CH2)10Br
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Quantity 100 g
Molecular Weight (g/mol) 235.209
Formula Weight 235.21
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-Bromoundecane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.