missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Catalog Number 10377700
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
112-71-0
C14H29Br
277.28
MFCD00000228
KOFZTCSTGIWCQG-UHFFFAOYSA-N
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
8208
1-bromotetradecane
CCCCCCCCCCCCCCBr
This item is not returnable. View return policy
112-71-0
C14H29Br
277.28
MFCD00000228
KOFZTCSTGIWCQG-UHFFFAOYSA-N
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
8208
1-bromotetradecane
CCCCCCCCCCCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 175.0°C to 178.0°C (20.0 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13Br
Refractive Index 1.4590 to 1.4610
Quantity 100 g
Beilstein 01,II,136
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 277.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'