missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopropane, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopropane |
| CAS | 106-94-5 |
| Molecular Formula | C3 H7 Br |
| Quantity | 500 g |
| Synonym | 1-Bromopropane, 1-Propyl bromide, 3-Bromopropane, Abzol JG, Ascusol MC, Drysolv, Leksol, PS Cleaner, Propyl bromide, n-Propyl bromide |
| InChI Formula | InChI=1S/C3H7Br/c1-2-3-4/h2-3H2,1H3 |
| SMILES | CCCBr |
| IUPAC Name | 1-bromopropane |
| Molecular Weight (g/mol) | 122.99 |
| Formula Weight | 121.9731 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction