missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluoroheptane, 97%

Product Code. 11449432
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11449432

Brand: Thermo Scientific Alfa Aesar A10156.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 375-88-2 | C7BrF15 | 448.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-88-2
Molecular Formula C7BrF15
Molecular Weight (g/mol) 448.96
MDL Number MFCD00013570
InChI Key VPQQZKWYZYVTMU-UHFFFAOYSA-N
Synonym 1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
PubChem CID 67819
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Density 1.894
Boiling Point 117°C to 119°C
Flash Point None
Linear Formula CF3(CF2)6Br
Refractive Index 1.303
Quantity 1 g
Beilstein 1716336
Formula Weight 448.96
Percent Purity 97%
Chemical Name or Material 1-Bromoperfluoroheptane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.