missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane, TRC

Product Code. 30328825
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328825 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328825 Supplier Toronto Research Chemicals Supplier No. TRCB68624525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromopentane
CAS 110-53-2
Molecular Formula C5H11Br
Quantity 25 g
Synonym 1-Bromopentane, 1-Pentyl Bromide, Amyl Bromide, NSC 7895, Pentyl Bromide, n-Amyl Bromide, n-Pentyl Bromide
InChI Formula InChI=1S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3
SMILES CCCCCBr
IUPAC Name 1-bromopentane
Molecular Weight (g/mol) 151.04
Formula Weight 150.0044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.