missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane-d11, TRC

Product Code. 30179427
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179427 1 g 1g
30179200 500 mg 500mg
30178946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179427 Supplier Toronto Research Chemicals Supplier No. TRCB6862471G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126840-21-9
Molecular Formula C5 D11 Br
Molecular Weight (g/mol) 162.113
Synonym Pentane-1,1,1,2,2,3,3,4,4,5,5-d11, 5-bromo- (9CI, ACI), 5-Bromopentane-1,1,1,2,2,3,3,4,4,5,5-d11 (ACI), 1-Bromo-1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentane, Pentyl-d11 bromide, 1-Bromopentane-d11, 1-Bromopentane-D11, 1-Bromopentane-1,1,2,2,3,3,4,4,5,5,5-d11
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentane
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])Br

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2
Formula Weight 161.073
Chemical Name or Material 1-Bromopentane-d11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.