missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane-d11, TRC

Product Code. 30179200
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179200 500 mg 500mg
30179427 1 g 1g
30178946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179200 Supplier Toronto Research Chemicals Supplier No. TRCB686247500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126840-21-9
Molecular Formula C5 D11 Br
Molecular Weight (g/mol) 162.113
Synonym Pentane-1,1,1,2,2,3,3,4,4,5,5-d11, 5-bromo- (9CI, ACI), 5-Bromopentane-1,1,1,2,2,3,3,4,4,5,5-d11 (ACI), 1-Bromo-1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentane, Pentyl-d11 bromide, 1-Bromopentane-d11, 1-Bromopentane-D11, 1-Bromopentane-1,1,2,2,3,3,4,4,5,5,5-d11
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentane
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])Br

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2
Formula Weight 161.073
Chemical Name or Material 1-Bromopentane-d11
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.