missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane, 99%

Product Code. 11489762
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489762 1000 g 1000g
11499762 250 g 250g
11409772 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489762 Supplier Thermo Scientific Alfa Aesar Supplier No. A10821.0B
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-53-2 | C5H11Br | 151.047 g/mol

1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.

Notes
Store away from oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

Melting Point -95°C
Density 1.215
Boiling Point 127°C to 129°C
Flash Point 31°C (87°F)
Odor Ether-like
Refractive Index 1.4436
Quantity 1000 g
UN Number UN1993
Beilstein 1730981
Merck Index 14,602
Solubility Information Insoluble in water and soluble in alcohol. Miscible in diethyl ether.
Formula Weight 151.05
Percent Purity 99%
Chemical Name or Material 1-Bromopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.