missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopentane, 98%

Product Code. p-76348 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10766161

Brand: Thermo Scientific Acros 152522500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-53-2 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -95°C
Color Colorless to Yellow
Density 1.2100g/mL
Boiling Point 130°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4Br
Refractive Index 1.4430 to 1.446
Quantity 250 mL
Beilstein 01, 131
Merck Index 15, 596
Specific Gravity 1.21
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,miscible with ether
Formula Weight 151.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromopentane, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromopentane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.