missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane, 98+%

Product Code. 11417756
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417756 250 g 250g
11407756 1000 g 1000g
11427756 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417756 Supplier Thermo Scientific Alfa Aesar Supplier No. A14722.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-83-1 | C8H17Br | 193.13 g/mol

1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr

Specifications

Melting Point -55°C
Density 1.114
Boiling Point 200°C to 202°C
Flash Point 78°C (172°F)
Odor Characteristic
Linear Formula CH3(CH2)7Br
Refractive Index 1.4518
Quantity 250 g
UN Number UN3082
Beilstein 1733136
Merck Index 14,6764
Solubility Information Immiscible with water.
Formula Weight 193.13
Percent Purity ≥98%
Chemical Name or Material 1-Bromooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.