missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane, 97%

Product Code. 11487427
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487427 100 g 100g
11497427 500 g 500g
11407437 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487427 Supplier Thermo Scientific Alfa Aesar Supplier No. A15588.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-89-0 | C18H37Br | 333.40 g/mol

1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.

Solubility
Soluble in acetone and most common organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-89-0
Molecular Formula C18H37Br
Molecular Weight (g/mol) 333.40
MDL Number MFCD00000231
InChI Key WSULSMOGMLRGKU-UHFFFAOYSA-N
Synonym octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
PubChem CID 8218
IUPAC Name 1-bromooctadecane
SMILES CCCCCCCCCCCCCCCCCCBr

Specifications

Melting Point 25°C to 30°C
Density 0.976
Boiling Point 150°C to 152°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Linear Formula CH3(CH2)17Br
Quantity 100 g
Beilstein 774145
Solubility Information Soluble in acetone and most common organic solvents. Insoluble in water.
Formula Weight 333.39
Percent Purity 97%
Chemical Name or Material 1-Bromooctadecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.