missing translation for 'onlineSavingsMsg'
Learn More

1-Bromononane, 99%

Product Code. 11443234
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443234 500 g 500g
11423234 25 g 25g
11433234 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443234 Supplier Thermo Scientific Alfa Aesar Supplier No. A10577.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-58-3 | C9H19Br | 207.155 g/mol

1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

Solubility
Insoluble in water. Soluble in ethyl ether and benzene.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-58-3
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00000278
InChI Key AYMUQTNXKPEMLM-UHFFFAOYSA-N
Synonym n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
PubChem CID 12742
IUPAC Name 1-bromononane
SMILES CCCCCCCCCBr

Specifications

Density 1.086
Boiling Point 200°C to 201°C
Flash Point 90°C (194°F)
Refractive Index 1.455
Quantity 500 g
Beilstein 1733632
Solubility Information Insoluble in water. Soluble in ethyl ether and benzene.
Formula Weight 207.16
Percent Purity 99%
Chemical Name or Material 1-Bromononane
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.