missing translation for 'onlineSavingsMsg'
Learn More

1-Bromononane, 97%

Product Code. 10050630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10050630 100 g Glass bottle 100g
10439590 25 g Glass bottle 25g
10299483 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10050630 Supplier Thermo Scientific Acros Supplier No. 107201000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-58-3 | C9H19Br | 207.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-58-3
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.15
MDL Number MFCD00000278
InChI Key AYMUQTNXKPEMLM-UHFFFAOYSA-N
Synonym n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
PubChem CID 12742
IUPAC Name 1-bromononane
SMILES CCCCCCCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0800g/mL
Boiling Point 201.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8Br
Refractive Index 1.4530 to 1.4550
Quantity 100 g
Beilstein 01, I, 63
Specific Gravity 1.08
Solubility Information Solubility in water: immiscible
Formula Weight 207.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromononane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromononane
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.