missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 97%, Thermo Scientific™

Product Code. 10280165
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10280165 5 g 5g
10344442 1 g 1g
10863761 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10280165 Supplier Thermo Scientific Maybridge Supplier No. SB01883DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 159689-88-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061271
InChI Key QSIVWRRHVXSDNE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-trifluoromethoxy benzene, 3-trifluoromethoxybenzyl bromide, alpha-bromo-3-trifluoromethoxy toluene, 3-trifluoromethoxy benzylbromide, m-trifluoromethoxy benzyl bromide, 3-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-3-trifluoromethoxy-benzene, benzene, 1-bromomethyl-3-trifluoromethoxy
PubChem CID 737176
IUPAC Name 1-(bromomethyl)-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(CBr)=C1

Especificaciones

CAS Min % 97.0
Density 1.5720g/mL
Assay Percent Range 97%
Linear Formula CH2BrC6H4OCF3
Quantity 5 g
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-3-(trifluoromethoxy)benzene
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.