missing translation for 'onlineSavingsMsg'
Learn More

1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™

Product Code. 10029142
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 82657-72-5
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
InChI Key YQRIQBOWLXRKKG-UHFFFAOYSA-N
Synonym 1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
PubChem CID 22675469
IUPAC Name 1-(bromomethyl)-2-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CBr

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-2-phenoxybenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'