missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoisoquinoline, 95+%, Thermo Scientific Chemicals

Product Code. 10415485
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1532-71-4
MDL Number MFCD00234478
InChI Key YWWZASFPWWPUBN-UHFFFAOYSA-N
Synonym isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888
PubChem CID 640963
IUPAC Name 1-bromoisoquinoline
SMILES C1=CC=C2C(=C1)C=CN=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H6BrN
Quantity 1 g
Molecular Weight (g/mol) 208.05
Formula Weight 208.05
Percent Purity 95+%
Physical Form Low Melting Solid
Chemical Name or Material 1-Bromoisoquinoline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromoisoquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'