missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoisoquinoline, 95+%

Product Code. 10320634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10320634 5 g Glass bottle 5g
10415485 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10320634 Supplier Thermo Scientific Supplier No. 392590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1532-71-4 | C9H6BrN | 208.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1532-71-4
MDL Number MFCD00234478
InChI Key YWWZASFPWWPUBN-UHFFFAOYSA-N
Synonym isoquinoline, 1-bromo, 1-bromo-isoquinoline, bromoisoquinoline, zlchem 702, pubchem6211, acmc-1bqnj, 1-bromoisoquinoline, abbypharma ap-12-10888
PubChem CID 640963
IUPAC Name 1-bromoisoquinoline
SMILES C1=CC=C2C(=C1)C=CN=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Molecular Formula C9H6BrN
Quantity 5 g
Molecular Weight (g/mol) 208.058
Formula Weight 208.05
Percent Purity ≥95%
Physical Form Low Melting Solid
Chemical Name or Material 1-Bromoisoquinoline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromoisoquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.