missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoisoquinolin-3-amine, 97%, Thermo Scientific™

Product Code. 10334562
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10334562 1 g Amber Glass Bottle 1g
10502462 250 mg Amber Glass Bottle 250mg
10673795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10334562 Supplier Thermo Scientific Maybridge Supplier No. BTB09943DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13130-79-5
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00102193
InChI Key PSQUIUNIVDKHJK-UHFFFAOYSA-N
Synonym 3-amino-1-bromoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromoisoquinolin-3-ylamine, 1-bromo-3-isoquinolylamine, 3-amino-1-bromo-isoquinoline, 1-bromoisoquinoline-3-amine, acmc-1by0f, 1-bromo-3-aminoisoquinoline, 1-bromanylisoquinolin-3-amine, 3-isoquinolinamine, 1-bromo
PubChem CID 289845
IUPAC Name 1-bromoisoquinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(N=C2Br)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-Bromoisoquinolin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.