missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexadecane, 98%

Product Code. 11422128
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422128 250 g 250g
11494180 25 g 25g
11412128 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422128 Supplier Thermo Scientific Alfa Aesar Supplier No. A15147.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-82-3 | C16H33Br | 305.34 g/mol

1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-82-3
Molecular Formula C16H33Br
Molecular Weight (g/mol) 305.34
MDL Number MFCD00000230
InChI Key HNTGIJLWHDPAFN-UHFFFAOYSA-N
Synonym hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
PubChem CID 8213
IUPAC Name 1-bromohexadecane
SMILES CCCCCCCCCCCCCCCCBr

Specifications

Melting Point 16°C to 18°C
Density 1
Boiling Point 336°C
Flash Point 176°C (348°F)
Odor Odorless
Refractive Index 1.462
Quantity 250 g
Beilstein 773989
Solubility Information Difficult to mix.
Formula Weight 305.35
Percent Purity 98%
Chemical Name or Material 1-Bromohexadecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.