missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, TRC

Product Code. 30337714
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30337714

Brand: Toronto Research Chemicals TRCB151411100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1-Bromoethyl)benzene
CAS 585-71-7
Molecular Formula C8H9Br
Quantity 100 mg
Synonym Benzene, (1-bromoethyl)- (6CI, 7CI, 8CI, 9CI, ACI), (1-Bromoethyl)benzene (ACI), (±)-(1-Bromoethyl)benzene, (±)-1-Bromo-1-phenylethane, (±)-1-Phenylethyl bromide, (α-Bromoethyl)benzene, 1-(1-Bromoethyl)benzene, 1-Bromo-1-phenylethane, 1-Phenethyl bromide, 1-Phenyl-1-bromoethane, 1-Phenylbromoethane, 1-Phenylethyl bromide, NSC 8052, α-Methylbenzyl bromide, α-Phenethyl bromide, α-Phenylethyl bromide
InChI Formula InChI=1S/C8H9Br/c1-7(9)8-5-3-2-4-6-8/h2-7H,1H3
SMILES CC(Br)c1ccccc1
IUPAC Name 1-bromoethylbenzene
Molecular Weight (g/mol) 185.06
Formula Weight 183.9888
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.