missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Bromoethyl)benzene, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (1-Bromoethyl)benzene |
| CAS | 585-71-7 |
| Molecular Formula | C8H9Br |
| Quantity | 1 g |
| Synonym | Benzene, (1-bromoethyl)- (6CI, 7CI, 8CI, 9CI, ACI), (1-Bromoethyl)benzene (ACI), (±)-(1-Bromoethyl)benzene, (±)-1-Bromo-1-phenylethane, (±)-1-Phenylethyl bromide, (α-Bromoethyl)benzene, 1-(1-Bromoethyl)benzene, 1-Bromo-1-phenylethane, 1-Phenethyl bromide, 1-Phenyl-1-bromoethane, 1-Phenylbromoethane, 1-Phenylethyl bromide, NSC 8052, α-Methylbenzyl bromide, α-Phenethyl bromide, α-Phenylethyl bromide |
| InChI Formula | InChI=1S/C8H9Br/c1-7(9)8-5-3-2-4-6-8/h2-7H,1H3 |
| SMILES | CC(Br)c1ccccc1 |
| IUPAC Name | 1-bromoethylbenzene |
| Molecular Weight (g/mol) | 185.06 |
| Formula Weight | 183.9888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction