missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoeicosane, TRC

Product Code. 30338136
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338136 Supplier Toronto Research Chemicals Supplier No. TRCB678838500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 1-Bromoeicosane
CAS 4276-49-7
Molecular Formula C20H41Br
Quantity 500 mg
Synonym 1-Bromoicosane, 1-Ecosyl Bromide, Eicosyl Bromide, NSC 87634, n-Eicosyl Bromide
InChI Formula InChI=1S/C20H41Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h2-20H2,1H3
SMILES CCCCCCCCCCCCCCCCCCCCBr
IUPAC Name 1-bromoicosane
Molecular Weight (g/mol) 361.44
Formula Weight 360.2392
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.