missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane, 98%

Product Code. 11403284
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403284 1000 g 1000g
11413284 250 g 250g
11423284 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403284 Supplier Thermo Scientific Alfa Aesar Supplier No. A10600.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143-15-7 | C12H25Br | 249.236 g/mol

1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

Melting Point -10°C
Density 1.04
Boiling Point 276°C
Flash Point 146°C (294°F)
Odor Weak
Linear Formula CH3(CH2)11Br
Refractive Index 1.458
Quantity 1000 g
Beilstein 506159
Merck Index 14,5389
Solubility Information Insoluble in water.
Formula Weight 249.24
Percent Purity 98%
Chemical Name or Material 1-Bromododecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.