missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromodecane, 98%
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
Description
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.
Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.
Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | -30°C |
| Density | 1.067 |
| Boiling Point | 238°C |
| Flash Point | 94°C (201°F) |
| Linear Formula | CH3(CH2)9Br |
| Refractive Index | 1.456 |
| Quantity | 500 g |
| UN Number | UN2810 |
| Beilstein | 1735227 |
| Solubility Information | Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water. |
| Show More |
Safety and Handling
Product Identifier
- 1-Bromodecane
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Bromodecane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction