missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoadamantane, 99%

Product Code. 11432697
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432697 100 g 100g
11422697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432697 Supplier Thermo Scientific Alfa Aesar Supplier No. A14893.22
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 768-90-1 | C10H15Br | 215.134 g/mol

1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

Solubility
It is soluble in organic solvents and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-90-1
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074721
InChI Key VQHPRVYDKRESCL-UHFFFAOYSA-N
Synonym 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane
PubChem CID 79106
IUPAC Name 1-bromoadamantane
SMILES C1C2CC3CC1CC(C2)(C3)Br

Specifications

Melting Point 117°C to 121°C
Quantity 100 g
Beilstein 1098857
Solubility Information It is soluble in organic solvents and insoluble in water.
Formula Weight 215.14
Percent Purity 99%
Chemical Name or Material 1-Bromoadamantane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.