missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-6-chlorohexane, 97%

Product Code. 11427244
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11427244

Brand: Thermo Scientific Alfa Aesar B21619.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6294-17-3 | C6H12BrCl | 199.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-17-3
Molecular Formula C6H12BrCl
Molecular Weight (g/mol) 199.52
MDL Number MFCD00001019
InChI Key JTYUIAOHIYZBPB-UHFFFAOYSA-N
Synonym 6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
PubChem CID 80516
IUPAC Name 1-bromo-6-chlorohexane
SMILES ClCCCCCCBr

Specifications

Density 1.337
Boiling Point 109°C to 110°C (2 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.4806
Quantity 25 g
Formula Weight 199.53
Percent Purity 97%
Chemical Name or Material 1-Bromo-6-chlorohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.