missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%

Product Code. 10470301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10470301 5 g Glass bottle 5g
10470491 25 g Glass bottle 25g
10003913 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10470301 Supplier Thermo Scientific Acros Supplier No. 221210050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4000g/mL
Boiling Point 210°C to 212°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Cl(CH2)5Br
Refractive Index 1.4826 to 1.4846
Quantity 5 g
Beilstein 01,III,346
Specific Gravity 1.4
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in methanol,ethanol,chloroform,and ether
Formula Weight 185.49
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-5-chloropentane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-5-chloropentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.