missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%

Product Code. 11424467
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424467 10 g 10g
11434467 50 g 50g
11444467 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424467 Supplier Thermo Scientific Alfa Aesar Supplier No. B20479.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol

1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr

Specifications

Density 1.408
Boiling Point 210°C to 212°C
Flash Point 95°C (203°F)
Refractive Index 1.4836
Quantity 10 g
Beilstein 1732341
Solubility Information Insoluble in water.
Formula Weight 185.5
Percent Purity 98%
Chemical Name or Material 1-Bromo-5-chloropentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.