missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethoxy)benzene, 98%

Product Code. 11480023
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480023 1 g 1g
11490023 5 g 5g
11400033 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480023 Supplier Thermo Scientific Alfa Aesar Supplier No. A12104.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 407-14-7 | C7H4BrF3O | 241.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-14-7
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD00040834
InChI Key SEAOBYFQWJFORM-UHFFFAOYSA-N
Synonym 1-bromo-4-trifluoromethoxy benzene, 4-trifluoromethoxy bromobenzene, 4-bromo trifluoromethoxy benzene, 4-trifluoromethoxybromobenzene, p-bromotrifluoromethoxybenzene, benzene, 1-bromo-4-trifluoromethoxy, 4-bromo-1-trifluoromethoxy benzene, p-trifluoromethoxybromobenzene, 4-bromotrifluoromethoxybenzene, 4-bromo-trifluoromethoxybenzene
PubChem CID 521008
IUPAC Name 1-bromo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(Br)C=C1

Specifications

Density 1.622
Boiling Point 155°C to 157°C
Flash Point 67°C (152°F)
Odor Unpleasant
Refractive Index 1.461
Quantity 1 g
Beilstein 2046332
Formula Weight 241.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4- (trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.