missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-tert-butylbenzene, 97%

Product Code. 11448466
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448466 500 g 500g
11428466 25 g 25g
11438466 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448466 Supplier Thermo Scientific Alfa Aesar Supplier No. A15924.36
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3972-65-4 | C10H13Br | 213.118 g/mol

1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

Melting Point 15°C to 16°C
Density 1.229
Boiling Point 231°C to 232°C
Flash Point 97°C (206°F)
Refractive Index 1.533
Quantity 500 g
Beilstein 1859117
Solubility Information Insoluble in water.
Formula Weight 213.12
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.