missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-pentylbenzene, 97%, ACROS Organics™

Product Code. 10527444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10527444 1 g 1g
10356444 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10527444 Supplier Thermo Scientific Acros Supplier No. 399460010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51554-95-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00061113
InChI Key SGCJPYYTVBHQGE-UHFFFAOYSA-N
Synonym 4-pentylbromobenzene, 4-bromo-n-pentylbenzene, 1-bromo-4-n-pentylbenzene, 4-n-amylbromobenzene, 4-n-pentylbromobenzene, 1-bromo-4-pentyl-benzene, p-bromopentylbenzene, 4-bromopentylbenzene, 1-amyl-4-bromobenzene, benzene, 1-bromo-4-pentyl
PubChem CID 2735599
IUPAC Name 1-bromo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)Br

Specifications

Assay Percent Range 97%
Linear Formula BrC6H4(CH2)4CH3
Quantity 1 g
Chemical Name or Material 1-Bromo-4-pentylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.