missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog Number 10664662
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
586-78-7
C6H4BrNO2
202.01
MFCD00007280
ZDFBKZUDCQQKAC-UHFFFAOYSA-N
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
11466
1-bromo-4-nitrobenzene
[O-][N+](=O)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
586-78-7
C6H4BrNO2
202.01
MFCD00007280
ZDFBKZUDCQQKAC-UHFFFAOYSA-N
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
11466
1-bromo-4-nitrobenzene
[O-][N+](=O)C1=CC=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 127.0°C
Color Yellow
Boiling Point 255.0°C to 256.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NO2
Quantity 10 g
Beilstein 05, 248
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 202.01
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-nitrobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-4-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'