missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-nitrobenzene, 98%

Product Code. 11434190
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434190 5 g 5g
11423288 25 g 25g
11433288 100 g 100g
11443288 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11434190 Supplier Thermo Scientific Alfa Aesar Supplier No. A17705.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol

It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-78-7
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007280
InChI Key ZDFBKZUDCQQKAC-UHFFFAOYSA-N
Synonym 4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
PubChem CID 11466
IUPAC Name 1-bromo-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 124°C to 127°C
Density 1.948
Boiling Point 254°C to 256°C
Quantity 5 g
UN Number UN3459
Beilstein 636964
Solubility Information Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.