missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-nonylbenzene, 98%

Product Code. 11466070
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466070 1 g 1g
11476070 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466070 Supplier Thermo Scientific Alfa Aesar Supplier No. H27042.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51554-94-0 | C15H23Br | 283.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51554-94-0
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.25
MDL Number MFCD00061116
InChI Key LVCWOFRWEVUMNY-UHFFFAOYSA-N
Synonym 1-bromo-4-n-nonylbenzene, bromononylbenzene, 4-nonylbromobenzene, 4-n-nonylbromobenzene, 4-nonylphenyl bromide, acmc-209kuh, 4-bromo-1-nonylbenzene, 1-bromo-4-nonylbenzene
PubChem CID 12603095
IUPAC Name 1-bromo-4-nonylbenzene
SMILES CCCCCCCCCC1=CC=C(Br)C=C1

Specifications

Density 1.115
Boiling Point 107°C (0.2 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.509
Quantity 1 g
Formula Weight 283.25
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-nonylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.