missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-hexyloxybenzene, 97%, Thermo Scientific Chemicals

Product Code. 11460581
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460581 5 g 5g
11450581 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460581 Supplier Thermo Scientific Alfa Aesar Supplier No. B20085.06
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30752-19-3
Molecular Formula C12H17BrO
Molecular Weight (g/mol) 257.171
MDL Number MFCD00173753
InChI Key GKLMJONYGGTHHM-UHFFFAOYSA-N
Synonym 1-bromo-4-hexyloxy benzene, 1-bromo-4-hexyloxybenzene, 1-4-bromophenoxy hexane, 1-bromo-4-n-hexyloxybenzene, 4-n-hexyloxybromobenzene, p-bromophenylhexyl ether, benzene, 1-bromo-4-hexyloxy, p-hexyloxybromobenzene, 4-hexyloxybromobenzene, 4-bromophenyl hexyl ether
PubChem CID 520492
IUPAC Name 1-bromo-4-hexoxybenzene
SMILES CCCCCCOC1=CC=C(C=C1)Br

Specifications

Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 5 g
Formula Weight 257.18
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-hexyloxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.