missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-isopropylbenzene, 97%

Product Code. p-7021148
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461003 5 g 5g
11359409 10 g 10g
11369409 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461003 Supplier Thermo Scientific Alfa Aesar Supplier No. A13370.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-61-8 | C9H11Br | 199.09 g/mol

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-61-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00039159
InChI Key MOZHUOIQYVYEPN-UHFFFAOYSA-N
Synonym 1-bromo-4-isopropylbenzene, 4-bromocumene, 4-bromoisopropylbenzene, p-bromocumene, 4-isopropylbromobenzene, benzene, 1-bromo-4-1-methylethyl, p-bromoisopropylbenzene, 2-p-bromophenyl propane, cumene, p-bromo, 2-4-bromophenyl propane
PubChem CID 11462
IUPAC Name 1-bromo-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(Br)C=C1

Specifications

Melting Point -12°C
Density 1.315
Boiling Point 219°C
Flash Point >100°C (212°F)
Refractive Index 1.536
Quantity 5 g
UN Number UN3082
Beilstein 1858096
Solubility Information Insoluble in water
Formula Weight 199.1
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-isopropylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.