missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-iodonaphthalene, 98%

Product Code. 15402209
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15402209

Brand: Thermo Scientific Alfa Aesar H64825.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 63279-58-3 | C10H6BrI | 332.966 g/mol

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Slightly soluble in water. (3.6 g/L) (25°C)

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63279-58-3
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.966
MDL Number MFCD01166242
InChI Key HQHHKYXPFKHLBF-UHFFFAOYSA-N
Synonym 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo
PubChem CID 5073667
IUPAC Name 1-bromo-4-iodonaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2I)Br

Specifications

Melting Point 84°C to 89°C
Quantity 250 mg
Beilstein 2044950
Sensitivity Light sensitive
Solubility Information Slightly soluble in water. (3.6g/L) (25°C)
Formula Weight 332.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Bromo-4-iodonaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.