missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoronaphthalene, 98%

Product Code. 11458766
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458766 25 g 25g
11444190 1 g 1g
11448766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458766 Supplier Thermo Scientific Alfa Aesar Supplier No. A16934.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 341-41-3 | C10H6BrF | 225.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341-41-3
Molecular Formula C10H6BrF
Molecular Weight (g/mol) 225.06
MDL Number MFCD00051473
InChI Key VAUJZKBFENPOCH-UHFFFAOYSA-N
Synonym 4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
PubChem CID 67647
IUPAC Name 1-bromo-4-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)F

Specifications

Melting Point 33°C to 37°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2085804
Formula Weight 225.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-fluoronaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.