missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethylbenzene, 99%, Thermo Scientific Chemicals

Catalog Number 11419923
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
1585-07-5
C8H9Br
185.064
MFCD00000111
URFPRAHGGBYNPW-UHFFFAOYSA-N
p-bromoethylbenzene, benzene, 1-bromo-4-ethyl, 4-ethylbromobenzene, p-ethylbromobenzene, 1-bromo-4-ethyl-benzene, unii-42li8tat6j, 1-ethyl-4-bromobenzene, 4-bromo-1-ethylbenzene, 4-ethyl-1-bromobenzene, 42li8tat6j
15313
1-bromo-4-ethylbenzene
CCC1=CC=C(C=C1)Br
This item is not returnable. View return policy
1585-07-5
C8H9Br
185.064
MFCD00000111
URFPRAHGGBYNPW-UHFFFAOYSA-N
p-bromoethylbenzene, benzene, 1-bromo-4-ethyl, 4-ethylbromobenzene, p-ethylbromobenzene, 1-bromo-4-ethyl-benzene, unii-42li8tat6j, 1-ethyl-4-bromobenzene, 4-bromo-1-ethylbenzene, 4-ethyl-1-bromobenzene, 42li8tat6j
15313
1-bromo-4-ethylbenzene
CCC1=CC=C(C=C1)Br

1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY
Density 1.341
Boiling Point 203°C to 204°C
Flash Point 63°C (145°F)
Refractive Index 1.544
Quantity 10 g
Beilstein 1856989
Solubility Information Insoluble in water.
Formula Weight 185.06
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-ethylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'