missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(dimethylamino)naphthalene, 95%

Product Code. 11328616
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11328616

Brand: Thermo Scientific Alfa Aesar L17666.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59557-93-6 | C12H12BrN | 250.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59557-93-6
Molecular Formula C12H12BrN
Molecular Weight (g/mol) 250.139
MDL Number MFCD02093945
InChI Key CWPDFSZHOZTGQW-UHFFFAOYSA-N
Synonym 1-bromo-4-dimethylamino naphthalene, acmc-20aox1, 4-bromonaphthyl dimethylamine, n,n-dimethyl-4-bromo-1-naphthylamine, n,n-dimethyl-4-bromonaphthalene-1-amine
PubChem CID 4083804
IUPAC Name 4-bromo-N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=CC=C(C2=CC=CC=C21)Br

Specifications

Density 1.4305
Boiling Point 137°C to 139°C (2 mmHg)
Refractive Index 1.6361
Quantity 1 g
UN Number UN2810
Beilstein 3254839
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material 1-Bromo-4-(dimethylamino)naphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.