missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(difluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10505280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10505280 5 g Glass bottle 5g
10822421 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10505280 Supplier Thermo Scientific Acros Supplier No. 432550050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51776-71-7
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD06657964
InChI Key HUSPSWKWFREKSS-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethyl benzene, 1-bromo-4-difluoromethyl-benzene, benzene, 1-bromo-4-difluoromethyl, 4-bromo-alpha,alpha-difluorotoluene, 4-bromobenzal fluoride, pubchem23296, acmc-1aukw, 1-bromo-4-difluoro-methyl-benzene
PubChem CID 2761183
IUPAC Name 1-bromo-4-(difluoromethyl)benzene
SMILES FC(F)C1=CC=C(Br)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 74.0°C to 76.0°C (12.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 207.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(difluoromethyl)benzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-4-(difluoromethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.