missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-butylbenzene, 97%

Product Code. 10239413 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239413 250 g Glass bottle 250g
10682752 10 g Glass bottle 10g
10400283 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239413 Supplier Thermo Scientific Acros Supplier No. 320442500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41492-05-1 | C10H13Br | 213.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41492-05-1
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00040934
InChI Key BRGVKVZXDWGJBX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
PubChem CID 521059
IUPAC Name 1-bromo-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4(CH2)3CH3
Refractive Index 1.5300 to 1.533
Quantity 250 g
Specific Gravity 1.2
Formula Weight 213.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-butylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.