missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-butylbenzene, 97%, Thermo Scientific Chemicals

Catalog Number 10400283
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
41492-05-1
C10H13Br
213.12
MFCD00040934
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
521059
1-bromo-4-butylbenzene
CCCCC1=CC=C(C=C1)Br
This item is not returnable. View return policy
41492-05-1
C10H13Br
213.12
MFCD00040934
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
521059
1-bromo-4-butylbenzene
CCCCC1=CC=C(C=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4(CH2)3CH3
Refractive Index 1.5300 to 1.533
Quantity 50 g
Specific Gravity 1.2
Formula Weight 213.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-butylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'