missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4,4,4-trifluorobutane, 98%

Product Code. p-7045314
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11378315 1 g 1g
11388315 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11378315

Brand: Thermo Scientific Alfa Aesar L10237.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 406-81-5 | C4H6BrF3 | 190.99 g/mol

1-Bromo-4,4,4-trifluorobutane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4,4,4-trifluorobutane is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406-81-5
Molecular Formula C4H6BrF3
Molecular Weight (g/mol) 190.99
MDL Number MFCD00066335
InChI Key DBCAQXHNJOFNGC-UHFFFAOYSA-N
Synonym 1-bromo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-bromobutane, 4,4,4-trifluorobutyl bromide, acmc-1ctt1, 4,4,4-trifluorobutylbromide, 1,1,1-trifluoro-4-bromobutane, 1-bromo 4,4,4-trifluoro butane, 4-bromo-1,1,1-trifluoro-butane, #, butane,4-bromo-1,1,1-trifluoro
PubChem CID 521006
IUPAC Name 4-bromo-1,1,1-trifluorobutane
SMILES FC(F)(F)CCCBr

Specifications

Density 1.555
Boiling Point 103°C to 105°C
Refractive Index 1.382
Quantity 1 g
Beilstein 1736058
Solubility Information Not miscible in water.
Formula Weight 191
Percent Purity 98%
Chemical Name or Material 1-Bromo-4,4,4-trifluorobutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.